missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Formamide, Spectrophotometric Grade, 99+%

Manufacturer:  Alfa Aesar™ 44066AP

 View more versions of this product

Catalog No. AA44066AP



Specifications

Specifications

Formamide
CH3NO
MFCD00007941
14,4237
Soluble in water (20mg/ml ),methanol,ethanol and acetone. Slightly soluble in ether.
C(=O)N
45.041
CHEBI:48431
Liquid
Spectrophotometric
Ammonia-like
154°C (310°F)
Colorless
500mL
75-12-7
HCONH2
505995
amid kyseliny mravenci
ZHNUHDYFZUAESO-UHFFFAOYSA-N
formamide
713
45.04
≥99%
1.134 g/mL
1.4475
210°C
2°C
Hygroscopic

Chemical Identifiers

75-12-7
45.041
ZHNUHDYFZUAESO-UHFFFAOYSA-N
713
formamide
CH3NO
MFCD00007941
amid kyseliny mravenci
CHEBI:48431
C(=O)N
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P260-P281-P308+P313-P501c

H351-H360FD-H373

EINECSNumber : 200-842-0

RTECSNumber : LQ0525000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents