missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Formamide, Molecular Biology Grade

Manufacturer:  Alfa Aesar™ J67520AE

 View more versions of this product

Catalog No. AAJ67520AE



Specifications

Specifications

Formamide
CH3NO
505995
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
C(=O)N
45.041
CHEBI:48431
Liquid
1.134g/mL
1.4475
Colorless
100mL
75-12-7
MFCD00007941
14,4237
ZHNUHDYFZUAESO-UHFFFAOYSA-N
formamide
713
45.04
Molecular Biology
Ammonia-like
210°C
2°C
Hygroscopic

Chemical Identifiers

75-12-7
45.041
ZHNUHDYFZUAESO-UHFFFAOYSA-N
713
formamide
CH3NO
MFCD00007941
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
CHEBI:48431
C(=O)N
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : LQ0525000

TSCA : TSCA

storageNote1 : 2° to 8°C

Documents