missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Formamide, Molecular Biology Grade

Manufacturer:  Alfa Aesar™ J67520AP

 View more versions of this product

Catalog No. AAJ67520AP



Specifications

Specifications

Formamide
CH3NO
505995
amid kyseliny mravenci
C(=O)N
45.041
CHEBI:48431
Liquid
1.134g/mL
1.4475
Colorless
500mL
75-12-7
MFCD00007941
14,4237
ZHNUHDYFZUAESO-UHFFFAOYSA-N
formamide
713
45.04
Molecular Biology
Ammonia-like
210°C
2°C
Hygroscopic

Chemical Identifiers

75-12-7
45.041
ZHNUHDYFZUAESO-UHFFFAOYSA-N
713
formamide
CH3NO
MFCD00007941
amid kyseliny mravenci
CHEBI:48431
C(=O)N
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : LQ0525000

TSCA : TSCA

storageNote1 : 2° to 8°C

Documents