Please login to your online account to display your discounted pricing

Formamide, Calbiochem™, EMD Millipore

Adenine deaminase inhibitor

$98.50 - $98.50

Chemical Identifiers

Chemical Name or Material Formamide
CAS 75-12-7
Molecular Formula CH3NO
Formula Weight 45
Synonym Carbamaldehyde, Methanamide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

344205500ML

 
emd millipore corporation
344205500ML
Each for $98.50
Description & Specifications

Specifications

Boiling Point 209.9°C
Melting Point 2.9°C
Formula Weight 45
Quantity 500mL
Chemical Name or Material Formamide
Odor Odorless
Protease None detected
Specific Gravity 1.1363
Storage Information Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
RNase None detected
Flash Point 154.9°C (310.8°F), Open Cup
Physical Form Liquid
Color Colorless
Molecular Formula CH3NO
CAS 75-12-7
Synonym Carbamaldehyde, Methanamide
Health Hazard 1 H360D May damage the unborn child.
Description Used in DNA stripping during genetic fingerprinting techniques.
DNase None detected
Safety and Handling

H360D May damage the unborn child.