missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formamide, ACS, 99.5+%, Alfa Aesar™

Manufacturer:  Alfa Aesar 14835D6

 View more versions of this product

Catalog No. AA14835D6



Specifications

Specifications

Formamide
75-12-7
CH3NO
MFCD00007941
14,4237
ZHNUHDYFZUAESO-UHFFFAOYSA-N
formamide
713
45.04
≥99.5%
1.134g/mL
1.4475
210°C
2°C
Hygroscopic
For ACS analysis
>99.5%
HCONH2
505995
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
C(=O)N
45.041
CHEBI:48431
Liquid
ACS
Ammonia-like
154° (310°F)
Colorless
4 x 1kg

Chemical Identifiers

75-12-7
45.041
ZHNUHDYFZUAESO-UHFFFAOYSA-N
713
formamide
CH3NO
MFCD00007941
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
CHEBI:48431
C(=O)N
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : LQ0525000

TSCA : TSCA

SDS Documents