Please login to your online account to display your discounted pricing

Formamide, 99.6%, ACS reagent, ACROS Organics™


Quantity 500g
Packaging EcoSafPak™ Glass Bottle

Manufacturer: thermo fisher scientific  423745000

 View more versions of this product

Catalog No. AC423745000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.5%
Purity Grade ACS Reagent Grade
Physical Form Liquid
Formula Weight 45.04
Quantity 500g
Color Clear
Beilstein 02, 26
Merck Index 15, 4265
Packaging EcoSafPak™ Glass Bottle
TSCA TSCA
EINECS Number 200-842-0
Melting Point 2.0° to 3.0°C
Density 1.56 (Air = 1.0)
RTECS Number LQ0525000
Fieser 01,402; 02,201; 03,147
Refractive Index 1.4460 to 1.4480 (20°C, 589nm)
Assay Percent Range 99.6%
Molecular Formula CH3NO
CAS 75-12-7
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May damage the unborn child.
Infrared Spectrum Authentic
Linear Formula HCONH2
MDL Number MFCD00007941
Chemical Name or Material Formamide
Additional Information Vapor Pressure: 0.08mbar at 20°C
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Formamide
CAS 75-12-7
Purity Grade ACS Reagent Grade
Assay ≥99.5%
Assay Percent Range 99.6%
Molecular Formula CH3NO
Linear Formula HCONH2
Formula Weight 45.04
MDL Number MFCD00007941
Accessories

Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May damage the unborn child.

SDS & Safety Documents