missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formamide, 99.5% min., ACS, EMD Millipore

Manufacturer:  Millipore Sigma FX04206

 View more versions of this product

Catalog No. MFX04206



Specifications

Specifications

Formamide
75-12-7
CH3NO
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
C(=O)N
45.041
CHEBI:48431
Slightly Viscous Liquid
1.13g/cm3 (20°C)
500°C
Colorless
For ACS analysis
99.5% min.
HCONH2
ZHNUHDYFZUAESO-UHFFFAOYSA-N
formamide
713
45.4
ACS/EP/Reagent
2.7 to 19.0%(V)
0.08 hPa (20°C)
1kg

Chemical Identifiers

75-12-7
45.041
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
CHEBI:48431
C(=O)N
CH3NO
ZHNUHDYFZUAESO-UHFFFAOYSA-N
713
formamide
Safety and Handling

Safety and Handling

Recommended Storage : Store at +15°C to +25°C

Documents