Please login to your online account to display your discounted pricing

Formamide, 99.5% min., ACS, EMD Millipore

$179.97 - $316.40

Chemical Identifiers

Chemical Name or Material Formamide
Name Note For ACS analysis
CAS 75-12-7
Purity Grade ACS/EP/Reagent Grade
Physical Form Slightly Viscous Liquid
Assay Percent Range 99.5% min.
Molecular Formula CH3NO
Linear Formula HCONH2
Formula Weight 45.4
Synonym Formic acid amide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MFX04206

 
emd millipore corporation
FX04206
1kg Each for $316.40

MFX04208

 
emd millipore corporation
FX04208
500g Each for $179.97
Description & Specifications

Specifications

Purity Grade ACS/EP/Reagent Grade
Physical Form Slightly Viscous Liquid
Formula Weight 45.4
Color Colorless
Quantity 1kg
Density 1.13g/cm3 (20°C)
Name Note For ACS analysis
Assay Percent Range 99.5% min.
Synonym Formic acid amide
Chemical Name or Material Formamide
Ignition Point 500°C
Vapor Pressure 0.08 hPa (20°C)
Molecular Formula CH3NO
Linear Formula HCONH2
CAS 75-12-7
Explosion Limit 2.7 to 19.0%(V)
Recommended Storage Store at +15°C to +25°C
Safety and Handling

Store at +15°C to +25°C