missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formamide, 99.5%, for analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 205821000

 View more versions of this product

Catalog No. AC205821000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
Specifications

Specifications

Formamide
75-12-7
210.0°C
175°C
Glass Bottle
1.4460 to 1.4480
1.133
HCONH2
3.75 mPa.s (20°C)
02, 26
15, 4265
Solubility in water: miscible with water. Other solubilities: miscible with methanol, ethanol, acetone, acetic, acid, dioxane, ethylene glycol, u.s.p. glycerol,, phenol, slibhtly soluble in ether and benzene
NC=O
45.04
CHEBI:48431
99.5%
2.0°C to 3.0°C
1.1330g/mL
100 mL
Authentic
Liquid
99.5%
CH3NO
MFCD00007941
0.05% max. (Coulometric)
01,402; 02,201; 03,147
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
ZHNUHDYFZUAESO-UHFFFAOYSA-N
formamide
713
45.04
Analytical

Chemical Identifiers

75-12-7
45.04
ZHNUHDYFZUAESO-UHFFFAOYSA-N
713
formamide
CH3NO
MFCD00007941
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
CHEBI:48431
NC=O
Safety and Handling

Safety and Handling

GHS P Statement May damage the unborn child. May cause damage to organs through prolonged or repeated exposure. Suspected of causing cancer if swallowed.

GHS P Statement Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Use personal protective equipment as required.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only