missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formamide, 99.5%, for analysis, ACROS Organics™

Manufacturer:  Acros Organics 205820025

 View more versions of this product

Catalog No. AC205820025



Description

Description

For analysis
Specifications

Specifications

Formamide
99.5%
HCONH2
02, 26
15, 4265
Solubility in water: miscible with water. Other solubilities: miscible with methanol, ethanol, acetone, acetic, acid, dioxane, ethylene glycol, u.s.p. glycerol,, phenol, slibhtly soluble in ether and benzene
C(=O)N
45.041
CHEBI:48431
Liquid
Analysis
1ppm max.
1ppm max.
1.4460 to 1.4480
1.133
0.05% max. (Coulometric)
3.75 mPa.s (20°C)
Undesignated
2.5L
75-12-7
CH3NO
MFCD00007941
01,402; 02,201; 03,147
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
ZHNUHDYFZUAESO-UHFFFAOYSA-N
formamide
713
45.04
99.5%
1.1330g/mL
Authentic
Glass bottle
Solubility in water: miscible with water. Other solubilities: miscible with methanol, ethanol, acetone, acetic, acid, dioxane, ethylene glycol, u.s.p. glycerol,, phenol, slibhtly soluble in ether and benzene
0.005% max.
175°C
210.0°C
2.0°C to 3.0°C

Chemical Identifiers

75-12-7
45.041
ZHNUHDYFZUAESO-UHFFFAOYSA-N
713
formamide
CH3NO
MFCD00007941
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
CHEBI:48431
C(=O)N
Safety and Handling

Safety and Handling

GHS P Statement May damage the unborn child. May cause damage to organs through prolonged or repeated exposure. Suspected of causing cancer if swallowed.

GHS P Statement Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Use personal protective equipment as required.

Danger

SDS Documents