Please login to your online account to display your discounted pricing

Formalin, HARLECO™, EMD Millipore


Quantity 4L

Manufacturer: emd millipore corporation  6534685

 View more versions of this product

Catalog No. M6534685

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 4L
Color Clear colorless
Chemical Name or Material Formalin
Name Note HARLECO™ Fixative
Physical Form Liquid
CAS 50-00-0

Specifications

Chemical Name or Material Formalin
Name Note HARLECO™ Fixative
CAS 50-00-0
Physical Form Liquid