Please login to your online account to display your discounted pricing

Formalin, HARLECO™, EMD Millipore

$96.34 - $890.03

Chemical Identifiers

Chemical Name or Material Formalin
Name Note HARLECO™ Fixative
CAS 50-00-0
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

M6534688

 
emd millipore corporation
6534688
20L Each for $281.54

M6511588

 
emd millipore corporation
6511588
20L Each for $890.03

M6534685

 
emd millipore corporation
6534685
4L Each for $96.34
Description & Specifications

Specifications

Quantity 20L
Color Clear colorless
Physical Form Liquid
Chemical Name or Material Formalin
Name Note HARLECO™ Fixative
CAS 50-00-0