Please login to your online account to display your discounted pricing

Formalin, 20%, EMD Millipore


Quantity 20L

Manufacturer: emd millipore corporation  R0458786

 View more versions of this product

Catalog No. MR0458786

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 20L
Color Clear
Physical Form Liquid
Chemical Name or Material Formalin, 20%
Synonym NBF Fixative
Assay Percent Range 20%

Specifications

Chemical Name or Material Formalin, 20%
Physical Form Liquid
Assay Percent Range 20%
Synonym NBF Fixative