Please login to your online account to display your discounted pricing

Formalin, 20%, EMD Millipore


Quantity 1cv

Manufacturer: emd millipore corporation  R0458701

 View more versions of this product

Catalog No. MR0458701

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1cv
Color Clear
Physical Form Liquid
Assay Percent Range 20%
Synonym NBF Fixative
Chemical Name or Material Formalin, 20%

Specifications

Chemical Name or Material Formalin, 20%
Physical Form Liquid
Assay Percent Range 20%
Synonym NBF Fixative