missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formalin, 20%, EMD Millipore

$538.57 - $580.69

Chemical Identifiers

Physical Form Liquid
Assay Percent Range 20%
Synonym NBF Fixative
Chemical Name or Material Formalin, 20%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MR0458786 Emd Millipore Corporation
R0458786
20L Each for $580.69
MR0458701 Emd Millipore Corporation
R0458701
1cv Each for $538.57
Specifications Description & Specifications

Chemical Identifiers

Liquid
20%
NBF Fixative
Formalin, 20%

Specifications

20L
20%
Clear
Liquid
NBF Fixative
Formalin, 20%