Please login to your online account to display your discounted pricing

Formalin, 10%, EMD Millipore

$127.23 - $2,119.69

Chemical Identifiers

Chemical Name or Material Formalin, 10%
Name Note Fixative
Physical Form Liquid
Assay Percent Range 10%
Synonym NBF Fixative
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MR0337982

 
emd millipore corporation
R0337982
4L Each for $228.49

MR0337986

 
emd millipore corporation
R0337986
20L Each for $494.51

MR0458682

 
emd millipore corporation
R0458682
4L Each for $179.49

MR0458684

 
emd millipore corporation
R0458684
10L Each for $306.06

MR0458686

 
emd millipore corporation
R0458686
20L Each for $406.94

MR0458676

 
emd millipore corporation
R0458676
1L Each for $127.23

MR0458690

 
emd millipore corporation
R0458690
200L Each for $2,119.69
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Chemical Name or Material Formalin, 10%
Name Note Fixative
Assay Percent Range 10%
Synonym NBF Fixative