Please login to your online account to display your discounted pricing

Formalin, 10%, EMD Millipore


Quantity 200L

Manufacturer: emd millipore corporation  R0458690

 View more versions of this product

Catalog No. MR0458690

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 200L
Chemical Name or Material Formalin, 10%
Name Note Fixative
Assay Percent Range 10%
Synonym NBF Fixative

Specifications

Chemical Name or Material Formalin, 10%
Name Note Fixative
Physical Form Liquid
Assay Percent Range 10%
Synonym NBF Fixative