Learn More
Formaldoxime trimer hydrochloride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2506318
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Formaldoxime trimer hydrochloride | |
∼128°C (decomposition) | |
MFCD00012778 | |
UN2811 | |
Hygroscopic | |
VRHJXINSVWQXEB-UHFFFAOYSA-N | |
1,3,5-trihydroxy-1,3,5-triazinane;hydrochloride | |
15919123 | |
95% |
6286-29-9 | |
C3H10ClN3O3 | |
50 g | |
3690722 | |
formaldoxime trimer hydrochloride, sym-triazinetriol hydrochloride, triformoxime hydrochloride, 1,3,5-triazinane-1,3,5-triol hydrochloride, 1,3,5-triazaperhydroine-1,3,5-triol, chloride, 1,3,5-triazine, hexahydro-1,3,5-trihydroxy-, hydrochloride | |
Cl.ON1CN(O)CN(O)C1 | |
171.58 | |
171.57 |
Chemical Identifiers
6286-29-9 | |
171.58 | |
VRHJXINSVWQXEB-UHFFFAOYSA-N | |
15919123 | |
Cl.ON1CN(O)CN(O)C1 |
C3H10ClN3O3 | |
MFCD00012778 | |
formaldoxime trimer hydrochloride, sym-triazinetriol hydrochloride, triformoxime hydrochloride, 1,3,5-triazinane-1,3,5-triol hydrochloride, 1,3,5-triazaperhydroine-1,3,5-triol, chloride, 1,3,5-triazine, hexahydro-1,3,5-trihydroxy-, hydrochloride | |
1,3,5-trihydroxy-1,3,5-triazinane;hydrochloride |
Safety and Handling
GHS H Statement
H300-H310-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
H300+H310-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 222-440-4
RTECSNumber : TQ0300000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only