missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Formaldehyde Solution, min. 37% GR for Analysis, ACS, Reagent Ph Eur Grade, MilliporeSigma™
Supplier: Technidata 1040031000
Description
For analysisSpecifications
Formaldehyde solution | |
50-00-0,67-56-1 | |
min. 37% | |
Plastic bottle | |
MFCD00003274 | |
UN2209 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde | |
712 | |
ACS/Reagent Ph. Eur. |
Stabilized with about 10% methanol | |
ca.300°C | |
Stinging | |
CH2O | |
1 L | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
C=O | |
30.03 | |
CHEBI:16842 | |
Liquid |
Chemical Identifiers
50-00-0 | |
30.03 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
712 | |
formaldehyde |
CH2O | |
MFCD00003274 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
CHEBI:16842 | |
C=O |
Safety and Handling
H227: Combustible liquid.
H301 + H311 + H331: Toxic if swallowed
DOTInformation : DOT Class: 8, Packaging Group: III