missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formaldehyde Solution, min. 37% Free From Acid, MilliporeSigma™

Manufacturer:  MilliporeSigma 1039991000

 View more versions of this product

Catalog No. M1039991000



Description

Description

For histology
Specifications

Specifications

Colorless liquid
ca.300°C
Plastic bottle
1L
50-00-0,67-56-1
CH2O
formalin
C=O
30.026
CHEBI:16842
Formaldehyde solution
Stinging
1.3 hPa
Stabilized with about 10% methanol and calcium carbonate
min. 37%
UN2209
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
712
Liquid

Chemical Identifiers

50-00-0
30.026
formalin
CHEBI:16842
C=O
CH2O
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
formaldehyde
Safety and Handling

Safety and Handling

H301 + H311 + H331: Toxic if swallowed

DOTInformation : DOT Class: 8, Packaging Group: III

Documents