Please login to your online account to display your discounted pricing

Formaldehyde Solution, ACS, Suitable for Histological Applications, Macron Fine Chemicals™

Suitable for histological applications

Chemical Identifiers

CAS 50-00-0
Purity Grade ACS Grade
Molecular Formula HCHO
Formula Weight 30.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK501602

 
avantor performance
501602
Poly Bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 30.03
Purity Grade ACS Grade
Quantity 500mL
Packaging Poly Bottle
Molecular Formula HCHO
CAS 50-00-0

macron