Please login to your online account to display your discounted pricing

Formaldehyde Solution ACS, EMD Millipore

$128.00 - $3,505.74

Chemical Identifiers

Chemical Name or Material Formaldehyde Solution
CAS 7732-18-5
Purity Grade ACS Grade
Linear Formula HCHO
Formula Weight 30.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MFX041020

 
emd millipore corporation
FX041020
Metal Drum w/bag 216kg Each for $3,505.74

MFX04105

 
emd millipore corporation
FX04105
Poly Bottle 450mL Each for $128.00
Description & Specifications

Specifications

Formula Weight 30.03
Purity Grade ACS Grade
Quantity 216kg
Packaging Metal Drum w/bag
Linear Formula HCHO
CAS 7732-18-5
Chemical Name or Material Formaldehyde Solution