missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Formaldehyde Solution, About 37%, MilliporeSigma™
Manufacturer: Emd Millipore Corporation 1040022500
Description
Recommended Storage
+15°C to +25°C.
Specifications
1.09 g/3 (20 °C) | |
93 to 96°C (1013 hPa) | |
>-15°C | |
2.8 to 4.0 (H2O, 20°C) | |
H5NO | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
azanium;hydroxide | |
14923 | |
Liquid |
Plastic Bottle | |
Colorless | |
2.5L | |
1336-21-6 | |
ammonia aqueous, ammonia water, ammonia, aqua, ammonia, monohydrate, ammonium hydroxide, ammonium hydroxide nh4 oh, ammonium hydroxide solution, aqua ammonia, aquammonia, household ammonia | |
[NH4+].[OH-] | |
35.046 | |
CHEBI:18219 | |
USP |
Chemical Identifiers
1336-21-6 | |
35.046 | |
ammonia aqueous, ammonia water, ammonia, aqua, ammonia, monohydrate, ammonium hydroxide, ammonium hydroxide nh4 oh, ammonium hydroxide solution, aqua ammonia, aquammonia, household ammonia | |
CHEBI:18219 | |
[NH4+].[OH-] |
H5NO | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
14923 | |
azanium;hydroxide |