Please login to your online account to display your discounted pricing

Formaldehyde-Sodium Carbonate Solution, Certified, LabChem™

$57.34 - $57.34

Chemical Identifiers

Chemical Name or Material Formaldehyde-Sodium Carbonate Solution
CAS 7732-18-5
CAS Max % 78.37
CAS 50-00-0
CAS Max % 16.45
CAS 67-56-1
CAS Max % 5.15
CAS 497-19-8
CAS Max % 0.03
Purity Grade Certified Grade
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC146801

 
lab chem
LC146801
500mL Each for $57.34
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 30.05
Quantity 500mL
Chemical Name or Material Formaldehyde-Sodium Carbonate Solution
CAS Max % 5.15
CAS 497-19-8
CAS Max % 0.03
Identification Passes Test
Physical Form Liquid
Packaging Poly Bottle
Linear Formula HCHO
CAS 7732-18-5
Solubility Soluble in water
Synonym Formalin
Traceability to NIST Traceable to NIST
Health Hazard 1 Danger
Decomposition Information Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Color Clear, colorless
pH 8.0 to 9.0
CAS Max % 78.37
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful if inhaled.
May cause cancer (inhalation).
Causes damage to organs (respiratory system, skin).
Harmful to aquatic life.
CAS 50-00-0
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing gas, mist, vapors, spray.
Wear protective clothing, protective gloves, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
If on skin: Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 16.45
Recommended Storage Room Temperature
CAS 67-56-1
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing gas, mist, vapors, spray.
Wear protective clothing, protective gloves, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
If on skin: Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful if inhaled.
May cause cancer (inhalation).
Causes damage to organs (respiratory system, skin).
Harmful to aquatic life.

Room Temperature