missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Formaldehyde, Buffered, Certified, 10% (v/v), 38 to 42g/L, LabChem™
Supplier: LabChem LC146705
Specifications
Formaldehyde | |
7732-18-5,50-00-0,67-56-1,7664-38-2,1310-73-2 | |
Phosphorus oxides; Carbon monoxide; Carbon dioxide | |
Cubitainer™ | |
20 L | |
30.05 | |
Certified |
Buffered | |
95,3.7,0.5,0.5,0.3 | |
Clear, colorless | |
HCHO | |
Formalin | |
10% (v/v) | |
Liquid |