Please login to your online account to display your discounted pricing

Formaldehyde, 37% by Weight (With Preservative/Certified ACS), Fisher Chemical


Stabilized with 10-15% methanol

Quantity 20L
Packaging PolyPac™

Manufacturer: fisher scientific company  F79P20

 View more versions of this product

Catalog No. F79P-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified ACS Grade
Formula Weight 30.03
Boiling Point 101°C
Melting Point 0°C
Quantity 20L
DOT Information DOT Class 3, : Flammable Liquid
Assay 36.5 to 38.0%
Physical Form Liquid
Color Colorless
Concentration or Composition (by Analyte or Components) Stabilized with 10-15% Methanol
Sulfate 0.002% max.
Methanol 9 to 15% v/v
Heavy Metals (as Pb) 5ppm max.
Synonym Formic Aldehyde
CAS 7732-18-5
ChemAlert Storage Symbol Red
MDL Number MFCD00003274
Packaging PolyPac™
Ignition Residue 0.005% max.
Chloride 5ppm max.
Iron (Fe) 2ppm max.
Assay Percent Range 36.5 to 38.0 %
Titratable Acid 0.006mEq/g max.
CAS 67-56-1
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes.

NFPA

Health:3
Flammability:2
Instability:0
CAS 50-00-0
Molecular Formula CH2O
Chemical Name or Material Formaldehyde, 37% by Weight
Linear Formula HCHO
Name Note With Preservative

Specifications

Chemical Name or Material Formaldehyde, 37% by Weight
Name Note With Preservative
CAS 50-00-0
CAS 67-56-1
CAS 7732-18-5
Purity Grade Certified ACS Grade
Assay 36.5 to 38.0%
Assay Percent Range 36.5 to 38.0 %
Molecular Formula CH2O
Linear Formula HCHO
Formula Weight 30.03
MDL Number MFCD00003274
Synonym Formic Aldehyde
Accessories

Accessories for Catalog Number: F79P20

Item Mfr. No. Item Description Price Per Unit Quantity  
296355103
Spilkleen/Cep  LAB SPILL KIT FORM/AMMONIA
spilkleen/cep
CSKLSK1FA
Each for $302.63
Accessories
 

Emergency Overview
Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes.

NFPA

Health:3
Flammability:2
Instability:0

DANGER!

SDS & Safety Documents