Please login to your online account to display your discounted pricing

Formaldehyde, 37% by Weight (With Preservative/Certified ACS), Fisher Chemical


Stabilized with 10-15% methanol

Quantity 1L
Packaging Glass Bottle

Manufacturer: fisher scientific company  F791

 View more versions of this product

Catalog No. F79-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified ACS Grade
Formula Weight 30.03
Boiling Point 101°C
Melting Point 0°C
Quantity 1L
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Assay 36.5 to 38.0%
Viscosity [NULL]
Color Colorless
Concentration or Composition (by Analyte or Components) Stabilized with 10-15% Methanol
Heavy Metals (as Pb) 5ppm max.
Iron (Fe) 2ppm max.
Ignition Residue 0.005% max.
Methanol 9 to 15% v/v
Assay Percent Range 36.5 to 38.0 %
MDL Number MFCD00003274
Sulfate 0.002% max.
Synonym Formic Aldehyde
Titratable Acid 0.006mEq/g max.
Chloride 5ppm max.
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes.

NFPA

Health:3
Flammability:2
Instability:0
CAS 7732-18-5
Chemical Name or Material Formaldehyde, 37% by Weight
Name Note With Preservative
Packaging Glass Bottle
Molecular Formula CH2O
Linear Formula HCHO
CAS 50-00-0
ChemAlert Storage Symbol Red
Health Hazard 2 DANGER!
CAS 67-56-1

Specifications

Chemical Name or Material Formaldehyde, 37% by Weight
Name Note With Preservative
CAS 50-00-0
CAS 67-56-1
CAS 7732-18-5
Purity Grade Certified ACS Grade
Assay 36.5 to 38.0%
Assay Percent Range 36.5 to 38.0 %
Molecular Formula CH2O
Linear Formula HCHO
Formula Weight 30.03
MDL Number MFCD00003274
Synonym Formic Aldehyde
Accessories

Accessories for Catalog Number: F791

Item Mfr. No. Item Description Price Per Unit Quantity  
Accessories
 

Emergency Overview
Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes.

NFPA

Health:3
Flammability:2
Instability:0

DANGER!

SDS & Safety Documents