missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 37%, ACS Reagent (Formalin), Reagents

Formaldehyde, 37%, ACS Reagent (Formalin), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

$116.62 - $1129.09

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Formalin, Methanal
IUPAC Name water
SMILES O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-337-956
View Documents
Labchem Inc
C120850010F
10 L
Each for $471.71
 
01-337-955
View Documents
Labchem Inc
C12085001A
1 L
Each for $155.97
 
01-337-954
View Documents
Labchem Inc
C120850020F
20 L
Each for $1,129.09
 
01-337-953
View Documents
Labchem Inc
C12085004A
4 L
Each for $284.35
 
01-337-952
View Documents
Labchem Inc
C1208500500A
500 mL
Each for $116.62
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
18.02
Formalin, Methanal
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water

Specifications

7732-18-5,50-00-0,67-56-1
96°C
H2O
1.073
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
ACS
Formaldehyde, 37%, ACS Reagent (Formalin)
Colorless
Cubitainer
10 L
Formalin, Methanal
O
18.02
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'