missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formaldehyde, 10% (v/v), Buffered, pH 7.4, Carson-Millonig Formulation, Ricca Chemical

Tissue Fixative for Light and Electron Microscopy

Manufacturer:  Ricca Chemical Company 31911

 View more versions of this product

Catalog No. 3191-1



Description

Description

Specifications

Specifications

4 L
Formaldehyde
7732-18-5,50-00-0,10049-21-5,1310-73-2
89.0,9.57,2.0,0.9
Buffered
Colorless
10% (v/v)
pH 7.4, Carson-Millonig Formulation
86.0,9.2,1.8,0.1
Natural Poly Bottle
Liquid
SDS
Documents
Product Certifications
Promotions

Promotions