Please login to your online account to display your discounted pricing

Formaldehyde, 10% (v/v) Buffered, pH 7.0, in Aqueous Phosphate Buffer, Ricca Chemical


Quantity 250mL
Packaging Poly natural

Manufacturer: ricca chemical company  RI3190250

 View more versions of this product

Catalog No. RIC01000BUF

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 250mL
Chemical Name or Material Formaldehyde
CAS 7732-18-5
CAS Max % 91.0
CAS Max % 9.74
Solvent or Matrix Aqueous Phosphate Buffer
CAS Min % 9.36
CAS Min % 87.0
CAS Min % 0.1
CAS Min % 0.1
Name Note Histological Fixative
CAS 50-00-0
CAS 10049-21-5
CAS 7558-79-4
Concentration or Composition (by Analyte or Components) 10% (v/v)
CAS Max % 0.9
Packaging Poly natural
CAS Max % 0.9

Specifications

Chemical Name or Material Formaldehyde
Name Note Histological Fixative
CAS 7732-18-5
CAS Min % 87.0
CAS Max % 91.0
CAS 50-00-0
CAS Min % 9.36
CAS Max % 9.74
CAS 10049-21-5
CAS Min % 0.1
CAS Max % 0.9
CAS 7558-79-4
CAS Min % 0.1
CAS Max % 0.9