Please login to your online account to display your discounted pricing

Formaldehyde, 10% (v/v) Aqueous Solution (non-buffered), Histological Fixative, Ricca Chemical

$90.33 - $90.33

Chemical Identifiers

Chemical Name or Material Formaldehyde, Non-buffered
Name Note Histological Fixative
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 92.0
CAS 50-00-0
CAS Min % 9.5
CAS Max % 9.88
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

3180-2-5

 
ricca chemical company
RI31802.5
Cubitainer 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3180-5

 
ricca chemical company
RI31805
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R3180000500

 
ricca chemical company
RIR3180000500A
Plastic Bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3180-1

 
ricca chemical company
RI31801
Poly natural 4L Each for $90.33
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 10L
Chemical Name or Material Formaldehyde, Non-buffered
Name Note Histological Fixative
Packaging Cubitainer
CAS 7732-18-5
CAS Min % 89.0
Concentration or Composition Notes 10% (v/v)
Solvent or Matrix Aqueous Solution
CAS Max % 92.0
CAS 50-00-0
CAS Min % 9.5
CAS Max % 9.88