missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formalde-Fresh Solution, Fisher Chemical

Manufacturer:  Fisher Scientific Company SF944

 View more versions of this product

Catalog No. SF94-4



 

Specifications Description & Specifications

Specifications

Formalde-Fresh Solution
50-00-0,67-56-1,7732-18-5
Pass Test
3.5 to 4.5%
Poly Bottle
93.9°C
Clear
4L
1.10g/cm³
Liquid
3.5 to 4.5 %
MFCD00003274
0°C
6.9 to 7.1

Chemical Identifiers

50-00-0,67-56-1,7732-18-5
MFCD00003274
3.5 to 4.5%
3.5 to 4.5 %
Formalde-Fresh Solution

Safety and Handling

SDS & Safety Documents

WARNING!

Emergency Overview
Cancer hazard. May cause an allergic skin reaction. Irritating to eyes, respiratory system and skin. May be harmful by inhalation, in contact with skin and if swallowed. May cause central nervous system effects. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Take precautionary measures against static discharges. Do not get in eyes, on skin, or on clothing. Do not breathe vapors or spray mist. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:2
Instability:0

DOTInformation : DOT Class 9: Miscellaneous