Please login to your online account to display your discounted pricing

Formalde-Fresh Solution, Fisher Chemical


Quantity 20L
Packaging PolyPac™

Manufacturer: fisher scientific company  SF9420

 View more versions of this product

Catalog No. SF94-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 93.9°C
Melting Point 0°C
Quantity 20L
DOT Information DOT Class 9: Miscellaneous
Density 1.10g/cm³
Physical Form Liquid
Assay 3.5 to 4.5%
Color Clear
pH 6.9 to 7.1
Chemical Name or Material Formalde-Fresh Solution
Packaging PolyPac™
MDL Number MFCD00003274
ChemAlert Storage Symbol Blue
Health Hazard 2 WARNING!
CAS 67-56-1
Health Hazard 3 Emergency Overview
Cancer hazard. May cause an allergic skin reaction. Irritating to eyes, respiratory system and skin. May be harmful by inhalation, in contact with skin and if swallowed. May cause central nervous system effects. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Take precautionary measures against static discharges. Do not get in eyes, on skin, or on clothing. Do not breathe vapors or spray mist. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:2
Instability:0
Identification Pass Test
Assay Percent Range 3.5 to 4.5 %
CAS 50-00-0
CAS 7732-18-5

Specifications

Chemical Name or Material Formalde-Fresh Solution
CAS 50-00-0
CAS 67-56-1
CAS 7732-18-5
Assay 3.5 to 4.5%
Assay Percent Range 3.5 to 4.5 %
MDL Number MFCD00003274
Accessories
 

Emergency Overview
Cancer hazard. May cause an allergic skin reaction. Irritating to eyes, respiratory system and skin. May be harmful by inhalation, in contact with skin and if swallowed. May cause central nervous system effects. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Take precautionary measures against static discharges. Do not get in eyes, on skin, or on clothing. Do not breathe vapors or spray mist. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:2
Instability:0

WARNING!

SDS & Safety Documents