missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formalde-Fresh Solution, Buffered, Fisher Chemical

Manufacturer:  Fisher Scientific Company SF9320

 View more versions of this product

Catalog No. SF93-20


 

Specifications Description & Specifications

Specifications

Formalde-Fresh Solution, Buffered
50-00-0,67-56-1,7732-18-5,7558-79-4
Liquid
PolyPac™
>1.00g/mol
Colorless
20L
Pass Test
4.0 to 4.5%
MFCD00003274
100°C
6.9 to 7.1

Chemical Identifiers

50-00-0,67-56-1,7732-18-5
MFCD00003274
Formalde-Fresh Solution, Buffered
4.0 to 4.5%

Safety and Handling

SDS & Safety Documents

WARNING!

Emergency Overview
Cancer hazard. May cause an allergic skin reaction. The toxicological properties have not been fully investigated. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:0

DOTInformation : DOT Class 9, : Miscellaneous