Innovating Science™ Forensic Chemistry of Blood Types Kit
Classify blood based on the presence or absence of erythrocytes on the surface of red blood cells.
Manufacturer: Innovating Science™ REFILLKI
Includes: Teacher's manual, student guide, analysis copy-masters, blood typing trays, four simulated blood samples (victim, suspect No. 1, suspect No. 2 and crime scene), ABO/Rh blood typing anti-sera, mixing sticks. Enough materials for 15 tests.
Description
Blood type, an inherited characteristic, is valuable to know in that it affects medical procedures, such as surgery and transfusions, paternity testing, as well as serving as evidence in criminal investigations.
Uses Innovating Science Simulated Blood; safe for classroom use.
Specifications
49 CFR 173.4 | |
White | |
14 | |
7 | |
Simulated blood samples, crime scene, abo/rh blood typing, anti-sera, Teacher's manual and student study guide copymasters | |
1 |
15 lab groups | |
20 | |
13 | |
For forensic chemistry of blood typing | |
Forensic Chemistry Of Blood Types Refill |