missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Fomblin YR-1800

$38.59 - $131.34

Chemical Identifiers

CAS 69991-67-9
MDL Number MFCD01323324
Synonym Perfluoropolyether YR-1800
Chemical Name or Material Fomblin YR-1800
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1754814 SDS Alfa Aesar™
L1754814
25g Each for $131.34
AAL1754806 SDS Alfa Aesar™
L1754806
5g Each for $38.59
Specifications Description & Specifications

Chemical Identifiers

69991-67-9
MFCD01323324
Perfluoropolyether YR-1800
Fomblin YR-1800

Specifications

69991-67-9
Perfluoropolyether YR-1800
Fomblin YR-1800
MFCD01323324
25g

Safety and Handling

TSCA : TSCA