Please login to your online account to display your discounted pricing

Alfa Aesar™ Fomblin YR-1800


Quantity 25g

Manufacturer: alfa aesar™  L1754814

 View more versions of this product

Catalog No. AAL1754814

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Fomblin YR-1800
TSCA TSCA
MDL Number MFCD01323324
CAS 69991-67-9
Synonym Perfluoropolyether YR-1800
Quantity 25g

Specifications

Chemical Name or Material Fomblin YR-1800
CAS 69991-67-9
MDL Number MFCD01323324
Synonym Perfluoropolyether YR-1800