missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Folinic acid, calcium salt pentahydrate, 95.0-105.0%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 230310010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Folinic acid, calcium salt pentahydrate | |
6035-45-6 | |
95% min. (HPLC) | |
C20H21CaN7O7·5H2O | |
1g | |
+ 14.30 | |
Solubility in water: freely soluble in water. Other solubilities: practically insoluble in alcohol | |
C1C(N(C2=C(N1)N=C(N=C2[O-])N)C=O)CNC3=CC=C(C=C3)C(=O)NC(CCC(=O)[O-])C(=O)O.O.O.O.O.O.[Ca+2] | |
601.58 | |
601.58 |
95.0-105.0% | |
Authentic | |
Glass bottle | |
MFCD00149465 | |
15, 4249 | |
folinic acid calcium pentahydrate | |
NPPBLUASYYNAIG-UHFFFAOYSA-L | |
calcium;4-[[4-[(2-amino-5-formyl-4-oxido-7,8-dihydro-6H-pteridin-6-yl)methylamino]benzoyl]amino]-5-hydroxy-5-oxopentanoate;pentahydrate | |
131674093 | |
95.0-105.0% |
Safety and Handling
AIR SENSITIVE
LIGHT SENSITIVE
EINECSNumber : 216-082-8
RTECSNumber : MA0600500
RUO â Research Use Only