missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Folic Acid, Fisher BioReagents

Manufacturer:  Fisher Scientific Company BP2519100

 View more versions of this product

Catalog No. BP2519100



 

Specifications Description & Specifications

Specifications

250°C
4.0 to 4.8
100g
C19H19N7O6
Yellow-Orange
0.3% max.
Pteroylglutamic Acid
≥95.0 %
253 to 259nm
Pass Test
Powder/Solid
441.39
15, 4248
1.59g/cm³
95.0% min.
59-30-3
Folic Acid
8.5% max.
Amber Glass
Conforms to reference

Chemical Identifiers

59-30-3
≥95.0 %
441.39
Folic Acid
Powder/Solid
C19H19N7O6
Pteroylglutamic Acid

Safety and Handling

SDS & Safety Documents

Recommended Storage : RT, protect from light