missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Folic Acid, Fisher BioReagents

$175.56 - $262.32

Chemical Identifiers

CAS 59-30-3
Molecular Formula C19H19N7O6
Molecular Weight (g/mol) 441.404
InChI Key OVBPIULPVIDEAO-LBPRGKRZSA-N
Synonym folacid
PubChem CID 6037
ChEBI CHEBI:27470
IUPAC Name (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP25195 SDS View Documents Fisher BioReagents
BP25195
5g Amber Glass Each for $203.29
Case of 7 Each for $1,423.03
 
BP251910 SDS View Documents Fisher BioReagents
BP251910
10g Amber Glass Each for $175.56
Case of 6 Each for $1,053.36
 
BP251925 SDS View Documents Fisher BioReagents
BP251925
25g Amber Glass Each for $262.32
Case of 3 Each for $786.96
 
Description

Description

Folic Acid is used in cell culture.
Specifications

Chemical Identifiers

59-30-3
441.404
folacid
CHEBI:27470
C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N
C19H19N7O6
OVBPIULPVIDEAO-LBPRGKRZSA-N
6037
(2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid

Specifications

1.59g/cm³
0.3% max.
Amber Glass
250°C
4.0 to 4.8
≥95.0 %
15, 4248
OVBPIULPVIDEAO-LBPRGKRZSA-N
(2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
6037
441.39
95.0% min.
Pass Test
8.5% max.
Yellow to Orange
5g
59-30-3
C19H19N7O6
folacid
C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N
441.404
CHEBI:27470
Powder/Solid
Folic Acid
Safety and Handling

Safety and Handling

Recommended Storage : RT, protect from light