Please login to your online account to display your discounted pricing

Foamtec UltraSORB Super Soft Foam Cleanroom Swabs

For removal of contamination from scratch sensitive optics

$117.76 - $117.76

Specifications

Adhesive Type Thermal
Product Type Swab
For Use With (Application) For removal of contamination from scratch sensitive optics
Handle Material Polypropylene
Shape Medium cylinder
View More Specs
Products
Catalog Number Mfr. No. Length (Metric) Length (English) Price Quantity    

19-304-626

 
foamtec
HT1411500
101.6mm 4 in. Pack of 500 Each for $117.76
Case of 16 Pack for $1,652.32
Description & Specifications

Specifications

Adhesive Type Thermal
Product Type Swab
For Use With (Application) For removal of contamination from scratch sensitive optics
Handle Material Polypropylene
Shape Medium cylinder
Tip Material CleanWIPE™ reticulated Polyurethane Foam
Tip Style Flexible

  • Non-scratching
  • No shedding
  • Solvent compatible
  • Long polypropylene handle
  • Super soft foam tip