Please login to your online account to display your discounted pricing
Foamtec ScrubCLEAN™ Abrasive Sponges
For cleaning hardened residues off scratch sensitive tools and components
$31.94 - $31.94
Specifications
Product Type Abrasive scrubber
Material Polyurethane Polymer
View More Specs
Products
Catalog Number Mfr. No. Style For Use With (Application) Price Quantity    
19-304-736
foamtec
FS206
Light duty Ink, paste, Epoxy removal from stencils and scrubbing bioreactors Pack of 2 Each for $31.94
Case of 50 Pack for $1,334.50
19-304-737
foamtec
FS208
Heavy duty Very hard deposition from shower heads, GDPs, CMP polishers Pack of 2 Each for $31.94
Case of 50 Pack for $1,334.50
Description & Specifications
Specifications
Product Type Abrasive scrubber
Material Polyurethane Polymer
For components such as CMP polishers, shower heads, heater surfaces, metal/polyester mesh masks, sealing surfaces, plastic and glass surfaces
  • Made of plastic particles that are adhered to sponge with polyurethane polymer
  • Minimizes particle generation from brasive surface
  • Low in endo-toxins, metals, salts and sodium
  • Autoclavable