Please login to your online account to display your discounted pricing

Foamtec PolyCLEAN™ Wipers

Lightweight, knitted-wiper constructed and processed to meet clean room requirements

$78.23 - $78.23

Specifications

Color Black
For Use With (Application) For particle sensitive wiping applications, ISO Class 5 and below clean room requirements
Product Type Polyclean™ cleanroom wipes
Material Polyester Fabric
View More Specs
Products
Catalog Number Mfr. No. Edges Width (English) Length (English) Price Quantity    

19-304-711

 
foamtec
HT5290L
Laser cut 9 in. 9 in. Pack of 150 Each for $78.23
Case of 10 Pack for $701.80
Description & Specifications

Specifications

Color Black
For Use With (Application) For particle sensitive wiping applications, ISO Class 5 and below clean room requirements
Product Type Polyclean™ cleanroom wipes
Material Polyester Fabric

Constructed from a continuous-filament fiber to resist shedding particles due to abrasion, making it the perfect choice for applications where cleanliness must be matched with cost control.

  • Washed and processed in a Class 10 environment making it ideal for particle sensitive wiping applications
  • Available in a black fabric for inspection and cleaning verification procedures
  • Amide and Silicone free for disk drive applications
  • Cut, washed and packed in our state-of-the-art integrated wiper facility to ensure product consistency