missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
FMOC-O-tert-Butyl-L-threonine, 98%, ACROS Organics™
Chemical Identifiers
CAS |
71989-35-0 |
Molecular Formula |
C23H27NO5 |
Molecular Weight (g/mol) |
397.45 |
MDL Number |
MFCD00077075 |
InChI Key |
LZOLWEQBVPVDPR-VLIAUNLRSA-N |
Synonym |
2s,3r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, fmoc-o-tert-butyl-l-threonine, fmoc-o-tert.butyl-l-threonine, fmoc-thr but, fmoc-thr but-oh, fmoc-thr otbu-oh, fmoc-thr t-bu-oh, fmoc-thr tbu-oh, n-9-fluorenylmethoxycarbonyl-o-tert-butyl-l-threonine, n-9h-fluoren-9-yl methoxy carbonyl-o-tert-butyl-l-threonine |
PubChem CID |
6364643 |
IUPAC Name |
(2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid |
SMILES |
CC(C(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)OC(C)(C)C |
View More Specs
Products
1
Catalog Number |
Mfr. No. |
Quantity |
Packaging |
Price |
Quantity |
|
|
|
|
AC265330010
SDS
View Documents
|
Thermo Fisher Scientific
265330010
|
1g |
Glass bottle |
N/A
|
N/A
|
|
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
|
|
Description
Specifications
Chemical Identifiers
|
71989-35-0 |
|
397.45 |
|
LZOLWEQBVPVDPR-VLIAUNLRSA-N |
|
6364643 |
|
CC(C(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)OC(C)(C)C |
|
C23H27NO5 |
|
MFCD00077075 |
|
2s,3r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, fmoc-o-tert-butyl-l-threonine, fmoc-o-tert.butyl-l-threonine, fmoc-thr but, fmoc-thr but-oh, fmoc-thr otbu-oh, fmoc-thr t-bu-oh, fmoc-thr tbu-oh, n-9-fluorenylmethoxycarbonyl-o-tert-butyl-l-threonine, n-9h-fluoren-9-yl methoxy carbonyl-o-tert-butyl-l-threonine |
|
(2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid |
Specifications
|
Authentic |
|
+ 16.00 (24.00°C c=1,EtOAc) |
|
White |
|
1g |
|
97.5% min. (HPLC) |
|
FMOC-NHCH(COOH)CH(CH3)OC(CH3)3 |
|
2s,3r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, fmoc-o-tert-butyl-l-threonine, fmoc-o-tert.butyl-l-threonine, fmoc-thr but, fmoc-thr but-oh, fmoc-thr otbu-oh, fmoc-thr t-bu-oh, fmoc-thr tbu-oh, n-9-fluorenylmethoxycarbonyl-o-tert-butyl-l-threonine, n-9h-fluoren-9-yl methoxy carbonyl-o-tert-butyl-l-threonine |
|
CC(C(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)OC(C)(C)C |
|
397.45 |
|
397.45 |
|
98% |
|
Glass bottle |
|
+ 16.00 |
|
131.0°C to 134.0°C |
|
71989-35-0 |
|
C23H27NO5 |
|
MFCD00077075 |
|
LZOLWEQBVPVDPR-VLIAUNLRSA-N |
|
(2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid |
|
6364643 |
|
Fine Fluffy Crystalline Powder |
|
FMOC-O-tert-Butyl-L-threonine |
Safety and Handling
SDS