missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
FMOC-O-tert-Butyl-L-threonine, 98%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific 265330010
Specifications
FMOC-O-tert-Butyl-L-threonine | |
White | |
97.5% min. (HPLC) | |
C23H26NO5 | |
MFCD00077075 | |
2s,3r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, fmoc-o-tert-butyl-l-threonine, fmoc-o-tert.butyl-l-threonine, fmoc-thr but, fmoc-thr but-oh, fmoc-thr otbu-oh, fmoc-thr t-bu-oh, fmoc-thr tbu-oh, n-9-fluorenylmethoxycarbonyl-o-tert-butyl-l-threonine, n-9h-fluoren-9-yl methoxy carbonyl-o-tert-butyl-l-threonine | |
LZOLWEQBVPVDPR-VLIAUNLRSA-M | |
+ 16.00 (24.00°C c=1,EtOAc) | |
+ 16.00 | |
6364643 | |
Fine Fluffy Crystalline Powder |
71989-35-0 | |
131.0°C to 134.0°C | |
Authentic | |
FMOC-NHCH(COOH)CH(CH3)OC(CH3)3 | |
Glass bottle | |
1g | |
C[C@@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O | |
(2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid | |
396.46 | |
397.45 | |
98% |
Chemical Identifiers
71989-35-0 | |
396.46 | |
LZOLWEQBVPVDPR-VLIAUNLRSA-M | |
6364643 | |
C[C@@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O |
C23H26NO5 | |
MFCD00077075 | |
2s,3r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, fmoc-o-tert-butyl-l-threonine, fmoc-o-tert.butyl-l-threonine, fmoc-thr but, fmoc-thr but-oh, fmoc-thr otbu-oh, fmoc-thr t-bu-oh, fmoc-thr tbu-oh, n-9-fluorenylmethoxycarbonyl-o-tert-butyl-l-threonine, n-9h-fluoren-9-yl methoxy carbonyl-o-tert-butyl-l-threonine | |
(2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid |
Safety and Handling
EINECSNumber : 276-261-1
RUO – Research Use Only