Please login to your online account to display your discounted pricing

FMOC-L-Norleucine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material FMOC-L-Norleucine
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C21H23NO4
Formula Weight 353.42
Synonym (S)-2-(FMOC-amino)hexanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC442720010

 
thermo fisher scientific
AC442720010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC442720050

 
thermo fisher scientific
AC442720050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 353.42
Quantity 1g
Color White to off-white
Melting Point 137° to 145°C
Molecular Formula C21H23NO4
CAS 77284-32-3
Synonym (S)-2-(FMOC-amino)hexanoic acid
Infrared Spectrum Authentic
Chemical Name or Material FMOC-L-Norleucine
Specific Rotation -18.0° to -20.0° (20°C, 589nm) (c-1, DMF)
Assay Percent Range 98%