missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FMOC-L-Methionine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 265310010

 View more versions of this product

Catalog No. AC265310010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

FMOC-L-Methionine
71989-28-1
C20H21NO4S
Authentic
2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, 9-fluorenylmethoxycarbonyl-l-methionine, fmoc-l-met-oh, fmoc-l-methionine, fmoc-met, fmoc-met-oh, methionine, n-9h-fluoren-9-ylmethoxy carbonyl, n-9h-fluoren-9-ylmethoxy carbonyl-l-methionine, n-alpha-9-fluorenylmethyloxycarbonyl-l-methionine, n-fmoc-l-methionine
BUBGAUHBELNDEW-SFHVURJKSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfanylbutanoic acid
2724632
-28° to -30° (24°C, 589nm) (c=1, DMF)
≥97.5%
White
98%
98%
FMOC-NHCH(COOH)CH2CH2SCH3
MFCD00037134
Glass bottle
CSCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
371.451
371.45
Granular Powder
−29.00 (24.00°C c=1,DMF)
1g

Chemical Identifiers

71989-28-1
371.451
BUBGAUHBELNDEW-SFHVURJKSA-N
2724632
CSCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C20H21NO4S
MFCD00037134
2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, 9-fluorenylmethoxycarbonyl-l-methionine, fmoc-l-met-oh, fmoc-l-methionine, fmoc-met, fmoc-met-oh, methionine, n-9h-fluoren-9-ylmethoxy carbonyl, n-9h-fluoren-9-ylmethoxy carbonyl-l-methionine, n-alpha-9-fluorenylmethyloxycarbonyl-l-methionine, n-fmoc-l-methionine
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfanylbutanoic acid
Safety and Handling

Safety and Handling

EINECSNumber : 276-258-5

SDS Documents