Please login to your online account to display your discounted pricing

FMOC-L-Methionine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material FMOC-L-Methionine
Name Note 98%
CAS 71989-28-1
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C20H21NO4S
Linear Formula FMOC-NHCH(COOH)CH2CH2SCH3
Formula Weight 371.45
MDL Number MFCD00037134
Synonym N-(9-Fluorenylmethoxycarbonyl)-L-methionine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC265310010

  SDS
thermo fisher scientific
265310010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 371.45
Physical Form Granular Powder
Assay ≥97.5%
Quantity 1g
Color White
EINECS Number 276-258-5
Specific Rotation Condition −29.00 (24.00°C c=1,DMF)
Synonym N-(9-Fluorenylmethoxycarbonyl)-L-methionine
Infrared Spectrum Authentic
Specific Rotation -28° to -30° (24°C, 589nm) (c=1, DMF)
Packaging Glass bottle
Chemical Name or Material FMOC-L-Methionine
Molecular Formula C20H21NO4S
CAS 71989-28-1
Name Note 98%
MDL Number MFCD00037134
Assay Percent Range 98%
Linear Formula FMOC-NHCH(COOH)CH2CH2SCH3