missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FMOC-L-arginine(PMC), 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC439870010

 View more versions of this product

Catalog No. AC439870010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(1mmol in 1mL DMF) clear, colorless
Authentic
-4.5
108°C to 109°C
119831-72-0
C35H42N4O7S
QTWZCODKTSUZJN-LJAQVGFWSA-N
(2S)-5-[[amino-[(2,2,5,7,8-pentamethyl-3,4-dihydrochromen-6-yl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
3010429
Powder
FMOC-L-Arginine(PMC)
Glass Bottle
White
1g
94% min. (HPLC)
ambotzfaa1351, fmoc-arg pmc, fmoc-arg pmc oh, fmoc-arg pmc-oh, fmoc-l-arg pmc-oh, n,a-fmoc-n, n-,a-fmoc-n-, o-2,2,5,7,8-pentamethylchroman, o-2,2,5,7,8-pentamethylchromane-6-sulfonyl-l-arginine, o-pmc-l-arginine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,5,7,8-pentamethylchroman-6-yl sulfonyl guanidino pentanoic acid
CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)N)C)CCC(O2)(C)C
662.81
662.81
95%

Chemical Identifiers

119831-72-0
662.81
ambotzfaa1351, fmoc-arg pmc, fmoc-arg pmc oh, fmoc-arg pmc-oh, fmoc-l-arg pmc-oh, n,a-fmoc-n, n-,a-fmoc-n-, o-2,2,5,7,8-pentamethylchroman, o-2,2,5,7,8-pentamethylchromane-6-sulfonyl-l-arginine, o-pmc-l-arginine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,5,7,8-pentamethylchroman-6-yl sulfonyl guanidino pentanoic acid
(2S)-5-[[amino-[(2,2,5,7,8-pentamethyl-3,4-dihydrochromen-6-yl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
C35H42N4O7S
QTWZCODKTSUZJN-LJAQVGFWSA-N
3010429
CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)N)C)CCC(O2)(C)C
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents