Please login to your online account to display your discounted pricing

FMOC-L-arginine(PMC), 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material FMOC-L-Arginine(PMC)
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C35H42N4O7S
Formula Weight 662.81
Synonym N-α-FMOC-N-ω-(2,2,5,7,8-pentamethylchromane-6-sulfonyl)-L-arginine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC439870010

  SDS
thermo fisher scientific
AC439870010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC439870050

  SDS
thermo fisher scientific
AC439870050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥94.0%
Formula Weight 662.81
Quantity 1g
Color White
Appearance of Solution (1mmol in 1mL DMF) clear, colorless
Synonym N-α-FMOC-N-ω-(2,2,5,7,8-pentamethylchromane-6-sulfonyl)-L-arginine
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C35H42N4O7S
CAS 119831-72-0
Chemical Name or Material FMOC-L-Arginine(PMC)
Specific Rotation -3.0° to -5.0° (20°C, 589nm) (C=1, DMF)
Assay Percent Range 95%
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.